Assay Details
Gene Symbol: LOC114114976 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970642.1 | XP_027826443.1 | 4 - 5 | 903 | 124 |
Gene Symbol: LOC101104490 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970644.1 | XP_027826445.1 | 2 - 3 | 344 | 124 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
