Assay Details
| Gene Symbol: ZNF445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012099476.2 | XP_011954866.1 | 4 - 5 | 883 | 84 | |
| XM_012099477.2 | XP_011954867.1 | 2 - 3 | 165 | 84 | ||
| XM_027958076.1 | XP_027813877.1 | 5 - 6 | 1002 | 84 | ||
More Information
Set Membership: |
|

ZNF445
Genomic Map