Assay Details
| Gene Symbol: MRPS18C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971146.1 | XP_027826947.1 | 3 - 4 | 285 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MRPS18C| Gene Symbol: MRPS18C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971146.1 | XP_027826947.1 | 3 - 4 | 285 | 91 | |
Set Membership: |
|