Assay Details
| Gene Symbol: CNNM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965918.1 | XP_027821719.1 | 4 - 5 | 1820 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CNNM3| Gene Symbol: CNNM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965918.1 | XP_027821719.1 | 4 - 5 | 1820 | 76 | |
Set Membership: |
|