Assay Details
| Gene Symbol: GK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021983.4 | XP_004022032.1 | 14 - 15 | 1265 | 88 | |
| XM_004021984.4 | XP_004022033.1 | 15 - 16 | 1283 | 88 | ||
| XM_012106441.3 | XP_011961831.1 | 16 - 17 | 1364 | 88 | ||
| XM_012106442.3 | XP_011961832.1 | 15 - 16 | 1345 | 88 | ||
| XM_012106443.3 | XP_011961833.1 | 16 - 17 | 1363 | 88 | ||
More Information
Set Membership: |
|

GK
Genomic Map