Assay Details
| Gene Symbol: LOC114113021 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964588.1 | XP_027820389.1 | 1 - 2 | 151 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC114113021| Gene Symbol: LOC114113021 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964588.1 | XP_027820389.1 | 1 - 2 | 151 | 98 | |
Set Membership: |
|