Assay Details
| Gene Symbol: SCAMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017847.4 | XP_004017896.1 | 4 - 5 | 450 | 89 | |
| XM_012098687.3 | XP_011954077.1 | 5 - 6 | 579 | 89 | ||
More Information
Set Membership: |
|

SCAMP2
Genomic Map