Assay Details
Gene Symbol: BARHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027968318.1 | XP_027824119.1 | 2 - 3 | 700 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
