Assay Details
| Gene Symbol: LOC101118831 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015092149.2 | XP_014947635.1 | 9 - 10 | 1374 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101118831| Gene Symbol: LOC101118831 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015092149.2 | XP_014947635.1 | 9 - 10 | 1374 | 113 | |
Set Membership: |
|