Assay Details
| Gene Symbol: ZBTB7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015095659.2 | XP_014951145.2 | 3 | 3179 | 108 | |
| XM_027969861.1 | XP_027825662.1 | 3 | 2970 | 108 | ||
| XM_027969862.1 | XP_027825663.1 | 3 | 3061 | 108 | ||
More Information
Set Membership: |
|

ZBTB7A
Genomic Map