Assay Details
| Gene Symbol: BASP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980150.1 | XP_027835951.1 | 2 | 740 | 91 | |
| XM_027980151.1 | XP_027835952.1 | 2 | 662 | 91 | ||
| XM_027980152.1 | XP_027835953.1 | 2 | 615 | 91 | ||
| XM_027980153.1 | XP_027835954.1 | 2 | 532 | 91 | ||
More Information
Set Membership: |
|

BASP1
Genomic Map