Assay Details
| Gene Symbol: LOC101104285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002524.4 | XP_004002573.1 | 3 | 386 | 101 | |
| Gene Symbol: LOC114113226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965191.1 | XP_027820992.1 | 3 | 358 | 101 | |
More Information
Set Membership: |
|

LOC101104285
Genomic Map