Assay Details
| Gene Symbol: BCDIN3D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004023317.3 | XP_004023366.3 | 1 | 357 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004023317.3 transcripts.
Set Membership: |
|

BCDIN3D
Genomic Map