Assay Details
| Gene Symbol: CANT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974213.1 | XP_027830014.1 | 4 | 1640 | 91 | |
| XM_027974214.1 | XP_027830015.1 | 5 | 1297 | 91 | ||
| XM_027974215.1 | XP_027830016.1 | 4 | 1189 | 91 | ||
More Information
Set Membership: |
|

CANT1
Genomic Map