Assay Details
| Gene Symbol: MGAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968128.1 | XP_027823929.1 | 2 | 1661 | 235 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGAT3| Gene Symbol: MGAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968128.1 | XP_027823929.1 | 2 | 1661 | 235 | |
Set Membership: |
|