Assay Details
| Gene Symbol: LOC101107365 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973513.1 | XP_027829314.1 | 1 | 1390 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101107365| Gene Symbol: LOC101107365 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973513.1 | XP_027829314.1 | 1 | 1390 | 70 | |
Set Membership: |
|