Assay Details
| Gene Symbol: LOC101110050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966008.1 | XP_027821809.1 | 1 | 4874 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101110050| Gene Symbol: LOC101110050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966008.1 | XP_027821809.1 | 1 | 4874 | 101 | |
Set Membership: |
|