Assay Details
| Gene Symbol: PAIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714019.3 | XP_002714065.1 | 9 - 10 | 1489 | 122 | |
| XM_008262175.2 | XP_008260397.1 | 9 - 10 | 1243 | 122 | ||
| XM_008262176.2 | XP_008260398.1 | 9 - 10 | 1137 | 122 | ||
| XM_017344711.1 | XP_017200200.1 | 10 - 11 | 1347 | 122 | ||
More Information
Set Membership: |
|

PAIP1
Genomic Map