Assay Details
| Gene Symbol: NEFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709311.4 | XP_002709357.3 | 1 - 2 | 1028 | 73 | |
| GenBank mRNA | Z47378.1 | - | 434 | 73 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NEFM| Gene Symbol: NEFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709311.4 | XP_002709357.3 | 1 - 2 | 1028 | 73 | |
| GenBank mRNA | Z47378.1 | - | 434 | 73 | ||
Set Membership: |
|