Assay Details
| Gene Symbol: UBE2D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717211.3 | XP_002717257.1 | 6 - 7 | 576 | 81 | |
| XM_008267547.2 | XP_008265769.1 | 6 - 7 | 577 | 81 | ||
| XM_008267548.2 | XP_008265770.1 | 6 - 7 | 576 | 81 | ||
| XM_008267549.2 | XP_008265771.1 | 6 - 7 | 526 | 81 | ||
More Information
Set Membership: |
|

UBE2D3
Genomic Map