Assay Details
Gene Symbol: ARCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008250462.2 | XP_008248684.1 | 605 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
