Assay Details
| Gene Symbol: EXTL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017337838.1 | XP_017193327.1 | 3 - 4 | 2424 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EXTL3| Gene Symbol: EXTL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017337838.1 | XP_017193327.1 | 3 - 4 | 2424 | 110 | |
Set Membership: |
|