Assay Details
| Gene Symbol: PCBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713231.3 | XP_002713277.1 | 12 - 13 | 1097 | 59 | |
| XM_017343857.1 | XP_017199346.1 | 11 - 12 | 1022 | 59 | ||
More Information
Set Membership: |
|

PCBP4
Genomic Map