Assay Details
| Gene Symbol: NDST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710310.3 | XP_002710356.1 | 10 - 11 | 2447 | 102 | |
| XM_008255264.2 | XP_008253486.1 | 10 - 11 | 2409 | 102 | ||
| XM_017341193.1 | XP_017196682.1 | 9 - 10 | 2265 | 102 | ||
More Information
Set Membership: |
|

NDST1
Genomic Map