Assay Details
| Gene Symbol: SH3BP5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723747.3 | XP_002723793.1 | 1222 | 65 | ||
| XM_017339831.1 | XP_017195320.1 | 2245 | 65 | |||
More Information
Set Membership: |
|

SH3BP5L
Genomic Map