Assay Details
| Gene Symbol: GRM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712087.3 | XP_002712133.1 | 3 - 4 | 1331 | 63 | |
| XM_008258240.2 | XP_008256462.1 | 3 - 4 | 1148 | 63 | ||
| XM_008258244.2 | XP_008256466.1 | 3 - 4 | 1225 | 63 | ||
More Information
Set Membership: |
|

GRM8
Genomic Map