Assay Details
| Gene Symbol: VPS37A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709510.3 | XP_002709556.1 | 7 - 8 | 950 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VPS37A| Gene Symbol: VPS37A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709510.3 | XP_002709556.1 | 7 - 8 | 950 | 110 | |
Set Membership: |
|