Assay Details
| Gene Symbol: ADNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008250426.2 | XP_008248648.1 | 211 | 144 | ||
| XM_008250427.2 | XP_008248649.1 | 1767 | 144 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADNP| Gene Symbol: ADNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008250426.2 | XP_008248648.1 | 211 | 144 | ||
| XM_008250427.2 | XP_008248649.1 | 1767 | 144 | |||
Set Membership: |
|