Assay Details
| Gene Symbol: CDH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257377.2 | XP_008255599.1 | 8 - 9 | 1883 | 100 | |
| XM_008257378.2 | XP_008255600.1 | 8 - 9 | 1882 | 100 | ||
| XM_008257379.2 | XP_008255601.1 | 8 - 9 | 1882 | 100 | ||
More Information
Set Membership: |
|

CDH8
Genomic Map