Assay Details
| Gene Symbol: XYLT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008271304.2 | XP_008269526.1 | 10 - 11 | 2371 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

XYLT2| Gene Symbol: XYLT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008271304.2 | XP_008269526.1 | 10 - 11 | 2371 | 83 | |
Set Membership: |
|