Assay Details
| Gene Symbol: PAX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718656.3 | XP_002718702.2 | 2 - 3 | 2502 | 61 | |
| XM_017348495.1 | XP_017203984.1 | 2 - 3 | 2502 | 61 | ||
| XM_017348496.1 | XP_017203985.1 | 2 - 3 | 2502 | 61 | ||
| XM_017348497.1 | XP_017203986.1 | 2 - 3 | 2502 | 61 | ||
More Information
Set Membership: |
|

PAX2
Genomic Map