Assay Details
| Gene Symbol: THRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719350.3 | XP_002719396.1 | 1 - 2 | 381 | 95 | |
| GenBank mRNA | EU489476.1 | - | 1 - 2 | 50 | 95 | |
| EU489477.1 | - | 1 - 2 | 50 | 95 | ||
More Information
Set Membership: |
|

THRA
Genomic Map