Assay Details
| Gene Symbol: MARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720913.3 | XP_002720959.1 | 1700 | 67 | ||
| XM_002720914.3 | XP_002720960.1 | 1699 | 67 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MARS1| Gene Symbol: MARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720913.3 | XP_002720959.1 | 1700 | 67 | ||
| XM_002720914.3 | XP_002720960.1 | 1699 | 67 | |||
Set Membership: |
|