Assay Details
| Gene Symbol: RBFOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017342437.1 | XP_017197926.1 | 9 - 10 | 1050 | 83 | |
| XM_017342438.1 | XP_017197927.1 | 1050 | 83 | |||
| XM_017342439.1 | XP_017197928.1 | 9 - 10 | 1050 | 83 | ||
More Information
Set Membership: |
|

RBFOX1
Genomic Map