Assay Details
| Gene Symbol: CPEB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710388.3 | XP_002710434.1 | 9 - 10 | 3391 | 67 | |
| XM_008255397.2 | XP_008253619.1 | 8 - 9 | 3367 | 67 | ||
| XM_008255398.2 | XP_008253620.1 | 8 - 9 | 3340 | 67 | ||
| XM_008255399.2 | XP_008253621.1 | 7 - 8 | 3316 | 67 | ||
More Information
Set Membership: |
|

CPEB4
Genomic Map