Assay Details
| Gene Symbol: YWHAE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718890.3 | XP_002718936.1 | 3 - 4 | 488 | 82 | |
| XM_008270937.2 | XP_008269159.1 | 3 - 4 | 490 | 82 | ||
More Information
Set Membership: |
|

YWHAE
Genomic Map