Assay Details
| Gene Symbol: GALT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272590.2 | XP_008270812.1 | 5 - 6 | 585 | 79 | |
| XM_017340724.1 | XP_017196213.1 | 5 - 6 | 532 | 79 | ||
| XM_017340727.1 | XP_017196216.1 | 5 - 6 | 586 | 79 | ||
More Information
Set Membership: |
|

GALT
Genomic Map