Assay Details
| Gene Symbol: PTEN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008270132.2 | XP_008268354.1 | 7 - 8 | 1056 | 91 | |
| XM_008270133.2 | XP_008268355.1 | 7 - 8 | 869 | 91 | ||
| XM_008270134.2 | XP_008268356.1 | 5 - 6 | 488 | 91 | ||
| XM_017348781.1 | XP_017204270.1 | 4 - 5 | 476 | 91 | ||
More Information
Set Membership: |
|

PTEN
Genomic Map