Assay Details
| Gene Symbol: PAIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710245.3 | XP_002710291.1 | 3 - 4 | 490 | 119 | |
| XM_008255100.2 | XP_008253322.1 | 3 - 4 | 469 | 119 | ||
More Information
Set Membership: |
|

PAIP2
Genomic Map