Assay Details
| Gene Symbol: FSD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708137.3 | XP_002708183.2 | 6 - 7 | 1167 | 82 | |
| XM_008257054.2 | XP_008255276.1 | 6 - 7 | 1170 | 82 | ||
| XM_008257148.2 | XP_008255370.1 | 5 - 6 | 1050 | 82 | ||
| XM_008257219.2 | XP_008255441.1 | 5 - 6 | 1039 | 82 | ||
More Information
Set Membership: |
|

FSD1L
Genomic Map