Assay Details
| Gene Symbol: WASHC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711226.3 | XP_002711272.1 | 6 - 7 | 590 | 91 | |
| XM_008256980.2 | XP_008255202.1 | 6 - 7 | 593 | 91 | ||
| XM_017342062.1 | XP_017197551.1 | 5 - 6 | 488 | 91 | ||
More Information
Set Membership: |
|

WASHC3
Genomic Map