Assay Details
| Gene Symbol: DHDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195678.1 | NP_001182607.1 | 787 | 79 | ||
| GenBank mRNA | AB021928.1 | - | 732 | 79 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DHDH| Gene Symbol: DHDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195678.1 | NP_001182607.1 | 787 | 79 | ||
| GenBank mRNA | AB021928.1 | - | 732 | 79 | ||
Set Membership: |
|