Assay Details
| Gene Symbol: HSP90B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256995.2 | XP_008255217.1 | 11 - 12 | 1489 | 78 | |
| GenBank mRNA | AF001631.1 | - | 9 - 10 | 1128 | 78 | |
More Information
Set Membership: |
|

HSP90B1
Genomic Map