Assay Details
| Gene Symbol: OXTR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195839.1 | NP_001182768.1 | 805 | 157 | ||
| XM_008273575.2 | XP_008271797.1 | 1282 | 157 | |||
| GenBank mRNA | AF023851.1 | - | 548 | 157 | ||
More Information
Set Membership: |
|

OXTR
Genomic Map