Assay Details
| Gene Symbol: OPN1MW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322264.1 | NP_001309193.1 | 431 | 75 | ||
| GenBank mRNA | AF031526.1 | - | 267 | 75 | ||
| DN891010.1 | - | 431 | 75 | |||
More Information
Set Membership: |
|

OPN1MW
Genomic Map