Assay Details
| Gene Symbol: HHEX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718509.3 | XP_002718555.1 | 3 - 4 | 668 | 93 | |
| GenBank mRNA | AF102133.1 | - | 2 - 3 | 232 | 93 | |
More Information
Set Membership: |
|

HHEX
Genomic Map