Assay Details
| Gene Symbol: ADAMTS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716775.3 | XP_002716821.1 | 4 - 5 | 1708 | 97 | |
| GenBank mRNA | AF317415.1 | - | 3 - 4 | 558 | 97 | |
More Information
Set Membership: |
|

ADAMTS5
Genomic Map