Assay Details
| Gene Symbol: PLPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714037.3 | XP_002714083.1 | 2 - 3 | 643 | 123 | |
| GenBank mRNA | AF404277.1 | - | 1 - 2 | 139 | 123 | |
More Information
Set Membership: |
|

PLPP1
Genomic Map