Assay Details
| Gene Symbol: SYNM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017337783.1 | XP_017193272.1 | 2452 | 145 | ||
| GenBank mRNA | AF510211.1 | - | 948 | 145 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SYNM| Gene Symbol: SYNM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017337783.1 | XP_017193272.1 | 2452 | 145 | ||
| GenBank mRNA | AF510211.1 | - | 948 | 145 | ||
Set Membership: |
|