Assay Details
| Gene Symbol: HAND2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249878.2 | XP_008248100.1 | 1 - 2 | 645 | 64 | |
| GenBank mRNA | AJ291308.1 | - | 1 - 2 | 345 | 64 | |
More Information
Set Membership: |
|

HAND2
Genomic Map